Skip to Content
Merck
All Photos(1)

Documents

A9402

Sigma-Aldrich

[Val5]-Angiotensin I bovine

≥97% (HPLC)

Synonym(s):

Asp-Arg-Val-Tyr-Val-His-Pro-Phe-His-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H87N17O14
CAS Number:
Molecular Weight:
1282.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1cnc[nH]1)NC(=O)C(Cc2ccccc2)NC(=O)C3CCCN3C(=O)C(Cc4cnc[nH]4)NC(=O)C(NC(=O)C(Cc5ccc(O)cc5)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CC(O)=O)C(C)C)C(C)C)C(O)=O

InChI

1S/C61H87N17O14/c1-32(2)22-46(60(91)92)75-54(85)44(25-37-28-65-30-68-37)71-53(84)42(23-35-12-8-7-9-13-35)72-56(87)47-15-11-21-78(47)59(90)45(26-38-29-66-31-69-38)74-58(89)50(34(5)6)77-55(86)43(24-36-16-18-39(79)19-17-36)73-57(88)49(33(3)4)76-52(83)41(14-10-20-67-61(63)64)70-51(82)40(62)27-48(80)81/h7-9,12-13,16-19,28-34,40-47,49-50,79H,10-11,14-15,20-27,62H2,1-6H3,(H,65,68)(H,66,69)(H,70,82)(H,71,84)(H,72,87)(H,73,88)(H,74,89)(H,75,85)(H,76,83)(H,77,86)(H,80,81)(H,91,92)(H4,63,64,67)

InChI key

HYGBEAAGLVPDGM-UHFFFAOYSA-N

Gene Information

cow ... AGT(527114)

Amino Acid Sequence

Asp-Arg-Val-Tyr-Val-His-Pro-Phe-His-Leu

Application

[Val5]-Angiotensin I is an Ang 1 peptide used in renin-angiotensin system studies.

Biochem/physiol Actions

Angiotensin I is a putative neurotransmitter. It is the precursor of angiotensin II and of angiotensin fragment 1-7, which are the hormones involved in regulation of fluid volume and the release of aldosterone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L G Navar et al.
American journal of hypertension, 13(1 Pt 2), 45S-54S (2000-03-11)
Because of the importance of the renin-angiotensin system in the pathophysiology of hypertension and in mediating associated alterations in renal function, angiotensin II (Ang II) AT1 receptor blockers provide a direct means of protecting against influences of excessive Ang II
S Kaneko et al.
Biochemical and biophysical research communications, 126(1), 587-593 (1985-01-16)
Two fractions with high potency for reversing the inhibitory effect of Met-enkephalin on the electrically induced contractility of guinea-pig ileum have been purified from bovine brain extract. Unexpectedly, one isolated peptide was identified as [Val5]-angiotensin I and the other fraction
Determination of renin activity in human plasma by column-switching high-performance liquid chromatography with fluorescence detection.
Miyazaki T, Kai M, Ohkura Y.
Journal of Chromatography A, 490, 43-51 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service