All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12N2O3
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98% (TLC)
form
powder
color
white
mp
268-270 °C
storage temp.
−20°C
SMILES string
CC(N)C(=O)NC(C)C(O)=O
InChI
1S/C6H12N2O3/c1-3(7)5(9)8-4(2)6(10)11/h3-4H,7H2,1-2H3,(H,8,9)(H,10,11)
InChI key
DEFJQIDDEAULHB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shifei Xia et al.
Electrophoresis, 30(16), 2837-2844 (2009-08-06)
Enantiomer of chiral dipeptides were separated by CE-ESI-MS in a bare fused-silica capillary using (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid (18C6H4) as the chiral selector. As 18C6H4 is a kind of nonvolatile chiral selector, in order to prevent from 18C6H4 into the ion-source of
Shahab A Shamsi
Electrophoresis, 23(22-23), 4036-4051 (2002-12-14)
A review is presented to highlight several approaches for coupling capillary electrophoresis (CE) and electrospray ionization-mass spectrometry (ESI-MS) for analysis of chiral compounds. A short discussion of commercially available CE-MS instruments and interface design is followed by a detail review
Ana Maria Castilla et al.
Beilstein journal of organic chemistry, 6, 5-5 (2010-05-21)
We present here the design, synthesis, and analysis of a series of receptors for peptide ligands inspired by the hydrogen-bonding pattern of protein β-sheets. The receptors themselves can be regarded as strands 1 and 3 of a three-stranded β-sheet, with
Matthias U Bohner et al.
The Journal of chemical physics, 137(3), 034105-034105 (2012-07-27)
The calculation of free-energy barriers by umbrella sampling and many other methods is hampered by the necessity for an a priori choice of the reaction coordinate along which to sample. We avoid this problem by providing a method to search
Delphine Patin et al.
Archives of microbiology, 194(6), 505-512 (2012-01-11)
Chlamydiaceae are obligate intracellular bacteria that do not synthesise detectable peptidoglycan although they possess an almost complete arsenal of genes encoding peptidoglycan biosynthetic activities. In this paper, the murF gene from Chlamydia trachomatis was shown to be capable of complementing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service