Skip to Content
Merck
All Photos(1)

Key Documents

A9103

Sigma-Aldrich

Anti-phospho-APP (pThr668) antibody produced in rabbit

affinity isolated antibody, aqueous glycerol solution, 10 blots

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

aqueous glycerol solution

usage

10 blots

mol wt

antigen 100-140 kDa

species reactivity

human

technique(s)

western blot: 1:1000 using CAD cells transfected with wild type vs. T668A mutant APP.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)
mouse ... App(11820)
rat ... App(54226)

General description

Amyloid precursor proteins (APPs) are transmembrane glycoproteins that are found in a wide range of tissues. APPs have 3 main isoforms, namely, APP695, APP751 and APP770 that are derived from alternative splicing events in cells. Accumulation of the cleavage products of APP, such as the β-amyloid peptide, can cause Alzheimer′s disease.
Rabbit Anti-phospho-APP (pThr668) antibody binds to human APP phosphorylated at Thr668. Mouse, rat and frog APP are 100% homologous.

Immunogen

synthetic phosphopeptide corresponding to the region of APP containing threonine 688.

Application

Cultured cortical neurons were immunostained for APP using rabbit anti-phospho-APP (pThr688) antibody.
Rabbit Anti-phospho-APP (pThr668) antibody can be used for western blot applications at a dilution of 1:1,000.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Roncarati et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(10), 7102-7107 (2002-05-16)
The beta-amyloid precursor protein (APP) and the Notch receptor undergo intramembranous proteolysis by the Presenilin-dependent gamma-secretase. The cleavage of APP by gamma-secretase releases amyloid-beta peptides, which have been implicated in the pathogenesis of Alzheimer's disease, and the APP intracellular domain
Chun-Yan Wang et al.
Antioxidants & redox signaling, 30(11), 1411-1431 (2018-04-11)
Oxidative stress and neuroinflammation play important roles in the pathology of Alzheimer's disease (AD). Thioredoxin-interacting protein (TXNIP), an endogenous inhibitor of antioxidant thioredoxin, is suspected to be an important modulator of oxidative stress and inflammation. However, the underlying mechanism involved
R E Tanzi et al.
Science (New York, N.Y.), 235(4791), 880-884 (1987-02-20)
The amyloid beta protein has been identified as an important component of both cerebrovascular amyloid and amyloid plaques of Alzheimer's disease and Down syndrome. A complementary DNA for the beta protein suggests that it derives from a larger protein expressed
R E Tanzi et al.
Nature, 331(6156), 528-530 (1988-02-11)
Amyloid B-protein/amyloid A4 is a peptide present in the neuritic plaques, neurofibrillary tangles and cerebrovascular deposits in patients with Alzheimer's disease and Down's syndrome (trisomy 21) and may be involved in the pathogenesis of Alzheimer's disease. Recent molecular genetic studies
D Goldgaber et al.
Science (New York, N.Y.), 235(4791), 877-880 (1987-02-20)
Four clones were isolated from an adult human brain complementary DNA library with an oligonucleotide probe corresponding to the first 20 amino acids of the beta peptide of brain amyloid from Alzheimer's disease. The open reading frame of the sequenced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service