Skip to Content
Merck
All Photos(1)

Key Documents

A8929

Sigma-Aldrich

Adrenocorticotropic Hormone Fragment 1-16 human, rat

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C89H133N25O22S
CAS Number:
Molecular Weight:
1937.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CO)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(N)CO)C(=O)NC(CCC(O)=O)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NCC(=O)NC(CCCCN)C(=O)N6CCCC6C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CCCCN)C(O)=O

Gene Information

human ... POMC(5443)
rat ... Pomc(24664)

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys

Other Notes

Fragment with weak adrenocorticotropic activity.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Bijlsma et al.
Neuroscience letters, 38(3), 297-302 (1983-08-08)
To measure protein synthesis at the level of the cell bodies of motoneurons of the sciatic nerve, anterior horns were punched out of the spinal cord using microdissection techniques and a cell-free system was prepared. Injections of ACTH1-16 and ACTH4-10
Heather Johnston et al.
Reproduction (Cambridge, England), 133(6), 1181-1187 (2007-07-20)
ACTH has been shown to stimulate androgen production by the fetal/neonatal mouse testis through the melanocortin type 2 receptor (MC2R). This study was designed to localize the expression of MC2R in the neonatal mouse testis and characterize the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service