Skip to Content
Merck
All Photos(1)

Key Documents

A8875

Sigma-Aldrich

N-Acetyl-DL-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO5
CAS Number:
Molecular Weight:
189.17
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CC(=O)NC(CCC(O)=O)C(O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily J Hart et al.
The Journal of biological chemistry, 284(9), 5977-5985 (2008-12-25)
Human carbamoyl phosphate synthetase (hCPS) has evolved critical features that allow it to remove excess and potentially neurotoxic ammonia via the urea cycle, including use of only free ammonia as a nitrogen donor, a K(m) for ammonia 100-fold lower than
Kazuyo Tujioka et al.
Journal of nutritional science and vitaminology, 51(2), 93-98 (2005-07-19)
The purpose of this study was to find whether the synthesis and degradation of N-acetylglutamate would affect urea synthesis when the dietary protein quality was manipulated. Experiments were done on three groups of rats given diets containing 10 g gluten
Xinying Jia et al.
Journal of biomolecular NMR, 44(2), 59-67 (2009-04-29)
Cell-free protein synthesis provides rapid and economical access to selectively 15N-labelled proteins, greatly facilitating the assignment of 15N-HSQC spectra. While the best yields are usually obtained with buffers containing high concentrations of potassium L-glutamate, preparation of selectively 15N-Glu labelled samples
Kazuyo Tujioka et al.
Journal of nutritional science and vitaminology, 57(1), 48-55 (2011-04-23)
We have shown that urinary urea excretion increased in rats given a lower quality protein. The purpose of present study was to determine whether the composition of dietary amino acids affects urea synthesis. Experiments were done on three groups of
Guangyan Qing et al.
Chirality, 21(3), 363-373 (2008-06-24)
Four linear thiourea anion receptors (1-3) derived from simple amino acid have been synthesized and their bonding properties with various chiral N-protected amino acid anions were examined by using UV-vis and fluorescence titration experiments. Receptors 1a, 2, and 3 exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service