Skip to Content
Merck
All Photos(1)

Key Documents

A8750

Sigma-Aldrich

N-Acetyl-β-D-glucosamine naphthol AS-LC

Synonym(s):

2-Acetamido-2-deoxy-β-D-glucosidoxynaphthol AS-LC, Naphthol AS-LC N-acetyl-β-D-glucosaminide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29ClN2O9
CAS Number:
Molecular Weight:
560.98
MDL number:
UNSPSC Code:
12171500

storage temp.

−20°C

SMILES string

COc1cc(NC(=O)c2cc3ccccc3cc2O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4NC(C)=O)c(OC)cc1C

Substrates

An interesting substrate for the histochemical demonstration of N-acetylglucosaminidase and for possible quantitative assay systems.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The localization of N-acetyl-beta-glucosaminidase in tissues.
D PUGH et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 9, 242-250 (1961-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service