Skip to Content
Merck
All Photos(1)

Documents

A8635

Sigma-Aldrich

1-Eicosanol

≥97%

Synonym(s):

Arachinyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19OH
CAS Number:
Molecular Weight:
298.55
Beilstein:
1705104
EC Number:
MDL number:
UNSPSC Code:
12352211

vapor density

7.4 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥97%

mp

62-65 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCO

InChI

1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3

InChI key

BTFJIXJJCSYFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

381.2 °F - open cup

Flash Point(C)

194 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Serafin et al.
The Journal of membrane biology, 248(4), 767-773 (2015-03-25)
Monolayers of phosphatidylcholine (PC), tetradecanol (TD), hexadecanol (HD), octadecanol (OD) and eicosanol (E) and their binary mixtures were investigated at the air/water interface. The surface tension values of pure and mixed monolayers were used to calculate π-A isotherms. The surface
Hajer Trabelsi et al.
Journal of the science of food and agriculture, 95(10), 2028-2032 (2014-09-13)
The search for other sources of vegetable oils by the exploitation and the enhancement of other oil plants will be needed to meet the demands of the international market. This study aims to determine the triacylglycerol (TAG) molecular species and
Giorgia Purcaro et al.
Analytical and bioanalytical chemistry, 407(1), 309-319 (2014-09-12)
The potentiality of a multidimensional comprehensive gas chromatographic (GC × GC) method, employing a simultaneous dual detection (FID and mass spectrometer), to generate peculiar two-dimensional chromatograms to be used as a chemical fingerprint, was investigated to characterize minor compounds in edible oil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service