Skip to Content
Merck
All Photos(1)

Key Documents

A8344

Sigma-Aldrich

Anti-ARC, C-Terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Apoptosis Repressor with CARD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 1:1,000 using HeLa cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOL3(8996)

General description

ARC is part of the activity regulated cytoskeleton-associated protein family. It is involved in endocytosis of AMPA receptors and formation of long-term memory. It is found expressed in the cytoplasma of neuronal dendrites.
ARC protein contains an N-terminal CARD that has homology to the CARDs present in Apaf-1 and RAIDD. It binds to and inhibits the activity of caspase 8. It also binds to caspase 9 and to Ced-3. It inhibits apoptosis induced by death receptors that require caspase 8 activation.
Rabbit Anti-ARC, C-Terminal antibody binds to amino acids 191-208 of human ARC.

Immunogen

synthetic peptide corresponding to amino acids 191-208 of human ARC (apoptosis repressor with CARD).

Application

Rabbit Anti-ARC, C-Terminal antibody can be used for western blot (1:1,000) and microarray analyses.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Koseki et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(9), 5156-5160 (1998-06-06)
We have identified and characterized ARC, apoptosis repressor with caspase recruitment domain (CARD). Sequence analysis revealed that ARC contains an N-terminal CARD fused to a C-terminal region rich in proline/glutamic acid residues. The CARD domain of ARC exhibited significant homology
Stefan J Kempf et al.
Oncotarget, 7(44), 71817-71832 (2016-10-07)
Accruing data indicate that radiation-induced consequences resemble pathologies of neurodegenerative diseases such as Alzheimer´s. The aim of this study was to elucidate the effect on hippocampus of chronic low-dose-rate radiation exposure (1 mGy/day or 20 mGy/day) given over 300 days

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service