Skip to Content
Merck
All Photos(1)

Key Documents

A7766

Sigma-Aldrich

N4-Acetylcytidine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N3O6
CAS Number:
Molecular Weight:
285.25
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98%

mp

199 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

CC(=O)NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C11H15N3O6/c1-5(16)12-7-2-3-14(11(19)13-7)10-9(18)8(17)6(4-15)20-10/h2-3,6,8-10,15,17-18H,4H2,1H3,(H,12,13,16,19)/t6-,8-,9-,10-/m1/s1

InChI key

NIDVTARKFBZMOT-PEBGCTIMSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kawai et al.
Nucleic acids symposium series, (21)(21), 61-62 (1989-01-01)
The conformational characteristics of a modified nucleoside 4-acetylcytidine (ac4C) was analyzed by proton nuclear magnetic resonance spectroscopy. This nucleoside was found to take the C3'-endo form predominantly. The enthalpy difference between the C2'-endo form and the C3'-endo form of the
Clement T Y Chan et al.
PLoS genetics, 6(12), e1001247-e1001247 (2010-12-29)
Decades of study have revealed more than 100 ribonucleoside structures incorporated as post-transcriptional modifications mainly in tRNA and rRNA, yet the larger functional dynamics of this conserved system are unclear. To this end, we developed a highly precise mass spectrometric
Yoshiho Ikeuchi et al.
The EMBO journal, 27(16), 2194-2203 (2008-08-01)
The wobble base of Escherichia coli elongator tRNA(Met) is modified to N(4)-acetylcytidine (ac(4)C), which is thought to ensure the precise recognition of the AUG codon by preventing misreading of near-cognate AUA codon. By employing genome-wide screen of uncharacterized genes in
Wilson R Sinclair et al.
ACS chemical biology, 12(12), 2922-2926 (2017-10-19)
The human acetyltransferase NAT10 has recently been shown to catalyze formation of N4-acetylcytidine (ac4C), a minor nucleobase known to alter RNA structure and function. In order to better understand the role of RNA acetyltransferases in biology and disease, here we
G S Morris et al.
Biomedical chromatography : BMC, 1(3), 109-118 (1986-06-01)
Inherited purine and pyrimidine disorders may be associated with serious, sometimes life-threatening consequences. Early and accurate diagnosis is essential. Difficulties encountered when using existing high pressure liquid chromatographic (HPLC) methods led to the development of an improved method based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service