Skip to Content
Merck
All Photos(1)

Key Documents

A4339

Sigma-Aldrich

N-Acetyl-Tyr-Val-Lys(biotinyl)-Asp 2,6-dimethylbenzoyloxymethyl ketone

≥90% (HPLC), powder

Synonym(s):

Ac-YVK(biotinyl)D 2,6-dimethylbenzoyloxymethyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€11.30

€11.30


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€11.30

About This Item

Empirical Formula (Hill Notation):
C46H63N7O12S
CAS Number:
Molecular Weight:
938.10
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

€11.30


Please contact Customer Service for Availability

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

DMF: 1 mg/mL

storage temp.

−20°C

Application

Useful for affinity labeling the larger subunit of activated caspase 1 and caspase 1 related proteases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dodheri Syed Samiulla et al.
Organic and medicinal chemistry letters, 2(1), 27-27 (2012-07-17)
Caspase-3 inhibition has been demonstrated to be therapeutically effective in moderating excessive programmed cell death. Interest in caspase-3 as a therapeutic target has led many to pursue the development of inhibitors. To date, only a few series of non-peptide inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service