Skip to Content
Merck
All Photos(1)

Key Documents

A3628

Sigma-Aldrich

Ala-Thr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4
CAS Number:
Molecular Weight:
190.20
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(C(C)N)=O)C(O)=O

InChI

1S/C7H14N2O4/c1-3(8)6(11)9-5(4(2)10)7(12)13/h3-5,10H,8H2,1-2H3,(H,9,11)(H,12,13)

InChI key

BUQICHWNXBIBOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Abe et al.
The Journal of general and applied microbiology, 65(1), 1-10 (2018-06-15)
The adenylation domain of nonribosomal peptide synthetase (NRPS) is responsible for its selective substrate recognition and activation of the substrate (yielding an acyl-O-AMP intermediate) on ATP consumption. DhbF is an NRPS involved in bacillibactin synthesis and consists of multiple domains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service