Skip to Content
Merck
All Photos(1)

Key Documents

A3105

Sigma-Aldrich

2-Amino-N-quinolin-8-yl-benzenesulfonamide

≥98% (HPLC), solid

Synonym(s):

QBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 22 mg/mL
H2O: insoluble

SMILES string

Nc1ccccc1S(=O)(=O)Nc2cccc3cccnc23

InChI

1S/C15H13N3O2S/c16-12-7-1-2-9-14(12)21(19,20)18-13-8-3-5-11-6-4-10-17-15(11)13/h1-10,18H,16H2

InChI key

NIOOKXAMJQVDGB-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of cell cycle at G2 phase; apoptosis inducer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Hee Kim et al.
Biochemical pharmacology, 69(9), 1333-1341 (2005-04-14)
We screened a library of 11,000 small molecular weight chemicals, looking for compounds that affect cell viability. We have identified 2-amino-N-quinoline-8-yl-benzenesulfonamide (QBS) as a potent cytotoxic compound that induces cell cycle arrest and apoptosis. Treatment of Jurkat T cells with
Cíntia Tiemi Misugi Kayukawa et al.
Food chemistry, 303, 125388-125388 (2019-08-28)
Saponins are known for their bioactive and surfactant properties, showing applicability to the food, cosmetic and pharmaceutical industries. This work evaluated the saponins effects on Kluyveromyces lactis β-galactosidase activity and correlated these changes to the protein structure. Enzyme kinetic was
Minji Kang et al.
ACS applied materials & interfaces, 9(3), 2686-2692 (2016-12-30)
We report on the fabrication of an organic thin-film semiconductor formed using a blend solution of soluble ambipolar small molecules and an insulating polymer binder that exhibits vertical phase separation and uniform film formation. The semiconductor thin films are produced
Fredy Omar Beltrán-Anaya et al.
Molecular oncology, 13(4), 909-927 (2019-01-17)
Triple negative breast cancer (TNBC) represents an aggressive phenotype with poor prognosis compared with ER, PR, and HER2-positive tumors. TNBC is a heterogeneous disease, and gene expression analysis has identified seven molecular subtypes. Accumulating evidence demonstrates that long non-coding RNA
Mónica Y Herrera-Sotero et al.
Nutrition and cancer, 72(5), 768-777 (2019-08-27)
Background: Breast and prostate cancer are frequently diagnosed neoplasias in women and men around the world. The signaling of the androgen receptor (AR) influences the development of both tumors. Since therapies focused to block the receptor's activity have not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service