Skip to Content
Merck
All Photos(1)

Key Documents

A277

Sigma-Aldrich

Monoclonal Anti-Na+/K+ ATPase (α1 Subunit) antibody produced in mouse

clone M8-P1-A3, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

M8-P1-A3, monoclonal

mol wt

antigen ~110 kDa

species reactivity

sheep, canine, rat, pig

should not react with

chicken, Xenopus

technique(s)

immunohistochemistry (frozen sections): 1:100
western blot: 1:250

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

Epitope mapping studies indicates the epitope is between amino acids 496 and 506 of lamb kidney Na+/K+ ATPase. No reactivity is observed in chicken or Xenopus. Staining is consistent with the plasma membrane localization of Na+/K+ ATPase.
Mouse monoclonal anti-Na+/K+ ATPase (α1 subunit) reacts with the α1 subunit of Na+/K+-ATPase in sheep, canine, rat and pig. In rat, the antibody shows no reactivity with the α2 or α3 subunits.
Na+/K+ transporting ATPase subunit α 1 is an ion transport pump critical for maintaining the gradient of Na and K ions across the plasma membrane.

Immunogen

lamb kidney α1 subunit of Na+/K+ ATPase.

Application

Mouse monoclonal anti-Na+/K+ ATPase (α1 subunit) has been used for western blot and co-immunoprecipitation assays. The product can also be used for immunohistochemistry applications using frozen sections at a dilution of 1:100.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen X Pu et al.
The Journal of biological chemistry, 277(23), 20270-20276 (2002-04-04)
The two variants of the gamma subunit of the rat renal sodium pump, gamma(a) and gamma(b), have similar effects on the Na,K-ATPase. Both increase the affinity for ATP due to a shift in the enzyme's E(1) <--> E(2) conformational equilibrium
Valdilene S Ribeiro et al.
Biochimica et biophysica acta, 1864(7), 2458-2471 (2018-04-15)
α-Tocopherol (α-Toc) overload increases the risk of dying in humans (E.R. Miller III et al. Meta-analysis: high-dosage vitamin E supplementation may increase all-cause mortality Ann Int Med. 142 (2005) 37-46), and overload during early development leads to elevation of blood
Mun Aw et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 314(4), R563-R573 (2018-01-20)
In general, the mammalian whole body mass-specific metabolic rate correlates positively with maximal urine concentration (Umax) irrespective of whether or not the species have adapted to arid or mesic habitat. Accordingly, we hypothesized that the thick ascending limb (TAL) of
W J Ball et al.
Biochimica et biophysica acta, 719(3), 413-423 (1982-12-17)
Four stable hybridoma cell lines secreting antibodies specific to the membrane (Na+ + K+)-dependent ATPase isolated from lamb kidney medulla have been produced by fusing mouse myeloma cells with spleen cells from immunized mice. These cell lines produce IgG gamma
Zinaeli López-González et al.
Journal of molecular histology, 51(6), 701-716 (2020-10-19)
The kidney controls body fluids, electrolyte and acid-base balance. Previously, we demonstrated that hyperpolarization-activated and cyclic nucleotide-gated (HCN) cation channels participate in ammonium excretion in the rat kidney. Since acid-base balance is closely linked to potassium metabolism, in the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service