Skip to Content
Merck
All Photos(1)

Key Documents

A270

Sigma-Aldrich

Anti-α1 Adrenergic Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ADRA1, Anti-ALPHA1BAR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse, human, rat

technique(s)

indirect immunofluorescence: 1:1,000 using mouse kidney distal tubule cells
western blot: 1:400

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Adrenergic receptors (ARs) are members of the 7-transmembrane domain G protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three α1AR subtypes (α1A/D, α1B, and α1C), three α2ARs (α2A, α2B, and α2C), and three βAR subtypes (β1, β2, and β3). Adrenergic receptors participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence.
Adrenergic receptors are a G-protien coupled receptors that exist in two forms, α or β. Ligands for adrenergic receptors are catecholamines that induce a sympathetic response in animal cells.
Rabbit polyclonal Anti-α1 Adrenergic Receptor antibody is human α1 adrenergic receptor subtype-specific and cross-reacts with rat and mouse α1 adrenergic receptor in immunoblotting. It detects α1 adrenergic receptor in mouse kidney membrane preparations by immunoblotting and stains plasma membrane of mouse kidney distal tubule cells in immunofluorescence.

Immunogen

A270 is produced by immunizing New Zealand white rabbits with a synthetic peptide corresponding to aa residues 339-349 of human alpha 1a adrenergic receptor. This sequence is 100% conserved within the 3rd intracellular loop of all alpha1 adrenergic receptors. Peptide sequence: K339-F-S-R-E-K-K-A-A-K-T349

Application

Rabbit polyclonal Anti-α1 Adrenergic Receptor antibody is used for the localization and detection of the α1 adrenergic receptor (~ 60 kDa). It can be used in immunoblotting and immunofluorescence techniques.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Roche et al.
Sub-cellular biochemistry, 72, 261-278 (2015-07-16)
Defining the physical-chemical determinants of protein folding and stability, under normal and pathological conditions has constituted a major subfield in biophysical chemistry for over 50 years. Although a great deal of progress has been made in recent years towards this
X D Tang et al.
The Journal of general physiology, 117(3), 253-274 (2001-02-27)
Reactive oxygen/nitrogen species are readily generated in vivo, playing roles in many physiological and pathological conditions, such as Alzheimer's disease and Parkinson's disease, by oxidatively modifying various proteins. Previous studies indicate that large conductance Ca(2+)-activated K(+) channels (BK(Ca) or Slo)
E S Drummond et al.
Neuroscience, 281, 99-109 (2014-10-01)
In certain forms of nerve injury and inflammation, noradrenaline augments pain via actions on up-regulated α1-adrenoceptors (α1-ARs). The aim of this study was to use immunohistochemistry to examine α1-AR expression on peripheral neurons, cutaneous blood vessels and keratinocytes after distal
Peter D Drummond et al.
Pain, 155(3), 606-616 (2013-12-18)
After peripheral nerve injury, nociceptive afferents acquire an abnormal excitability to adrenergic agents, possibly due to an enhanced expression of α1-adrenoceptors (α1-ARs) on these nerve fibres. To investigate this in the present study, changes in α1-AR expression on nerve fibres
Mária Dux et al.
European journal of pain (London, England), 24(2), 383-397 (2019-10-30)
The α1 -adrenoceptor agonist, phenylephrine, is used at high concentrations as a mydriatic agent and for the treatment of nasal congestion. Among its adverse side-effects transient burning sensations are reported indicating activation of the trigeminal nociceptive system. Neuropeptide release, calcium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service