Skip to Content
Merck
All Photos(1)

Documents

A2503

Sigma-Aldrich

DL-Alanine β-naphthylamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CONHC10H7·HCl
CAS Number:
Molecular Weight:
250.72
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.83

Assay

≥98% (TLC)

form

powder

mp

258-260 °C (dec.) (lit.)

solubility

ethanol: 50 mg/mL, clear to slightly hazy

storage temp.

2-8°C

SMILES string

Cl.CC(N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C13H14N2O.ClH/c1-9(14)13(16)15-12-7-6-10-4-2-3-5-11(10)8-12;/h2-9H,14H2,1H3,(H,15,16);1H

InChI key

WNLRRMRLNYQNOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-alanine β-naphthylamide (DLABN) has been used as a substrate to treat Listeria in the hydrolysis test to compare methods for the identification of Listeria species. It has been used as a substrate in the hydrolysis of DLABN to differentiate Listeria monocytogenes from other Listeria species.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Buckler et al.
Biochemistry, 34(49), 15965-15978 (1995-12-12)
Formation of local structure and overall chain dimensions in the 124-residue, four-disulfide protein bovine pancreatic ribonuclease A (RNase A) under conditions favoring either the native or partially folded states have been studied by nonradiative excitation energy transfer measurements. Three RNase
C I Cheeseman et al.
Canadian journal of physiology and pharmacology, 60(9), 1177-1184 (1982-09-01)
The uptake of the peptide glycyl-L-leucine across the brush border of the rat small intestinal enterocyte was studied using everted rings. The transfer of leucine from the dipeptide into the enterocyte was greater than the glycine uptake from glycyl-L-leucine. This
A G Clark et al.
Journal of clinical microbiology, 35(8), 2155-2156 (1997-08-01)
The hydrolysis of DL-alanine-beta-naphthylamide and D-alanine-p-nitroanilide for identification of Listeria spp. has been studied with 227 cultures. All species of Listeria, except L. monocytogenes, hydrolyzed these substrates. The reactions were detected by simple chromogenic reactions and could substitute for the
P Kugler et al.
Histochemistry, 82(4), 397-400 (1985-01-01)
The localization of exopeptidase activities was demonstrated histochemically (by simultaneous azo coupling) on the visceral endoderm of whole unfixed yolk sacs of rats (12.5-18.5 days of gestation). For comparison, the topochemistry of exopeptidases was studied by conventional section histochemistry of
J McLauchlin
International journal of food microbiology, 38(1), 77-81 (1997-08-19)
The purpose of this study was to compare methods for the identification of Listeria species. Three hundred and fifty cultures representing the six species of Listeria were tested using conventional sugar fermentation and haemolytic reactions, as well as the hydrolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service