Skip to Content
Merck
All Photos(1)

Key Documents

A242

Sigma-Aldrich

Alloxazine

solid

Synonym(s):

Benzo[g]pteridine-2,4(1H,3H)-dione, Isoalloxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N4O2
CAS Number:
Molecular Weight:
214.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

color

yellow

solubility

DMSO: 11 mg/mL
dilute aqueous base: 4 mg/mL
H2O: insoluble
dilute aqueous acid: insoluble
ethanol: insoluble

SMILES string

O=C1NC(=O)c2nc3ccccc3nc2N1

InChI

1S/C10H6N4O2/c15-9-7-8(13-10(16)14-9)12-6-4-2-1-3-5(6)11-7/h1-4H,(H2,12,13,14,15,16)

InChI key

HAUGRYOERYOXHX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective A2B adenosine receptor antagonist.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - Solvent

Flash Point(C)

87 °C - Solvent


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B T Liang et al.
Circulation research, 76(2), 242-251 (1995-02-01)
Adenosine exerts pronounced biological effects in the heart cell. The role of multiple adenosine receptor subtypes in regulating the heart cell function is not known. Ventricular cells cultured from chick embryos 14 days in ovo were used to study a
L E Brackett et al.
Biochemical pharmacology, 47(5), 801-814 (1994-03-02)
The adenosine (ADO) receptor in NIH 3T3 fibroblasts was characterized using a series of adenosine agonists and selected xanthine and non-xanthine antagonists. The ADO receptor elicited accumulations of cyclic AMP in intact NIH 3T3 fibroblasts and caused activation of adenylate
Patrizia Minetti et al.
Journal of medicinal chemistry, 48(22), 6887-6896 (2005-10-28)
Two types of adenosine receptor ligands were designed, i.e., 9H-purine and 1H-imidazo[4,5-c]pyridines, to obtain selective A(2A) antagonists, and we report here their synthesis and binding affinities for the four adenosine receptor subtypes A(1), A(2A), A(2B) and A(3). The design was
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service