Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C9H19N3O3 · HCl
CAS Number:
Molecular Weight:
253.73
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Product Name
Ala-Lys hydrochloride,
assay
≥98% (TLC)
form
powder
color
white to off-white
application(s)
peptide synthesis
storage temp.
−20°C
SMILES string
Cl.CC(N)C(=O)NC(CCCCN)C(O)=O
InChI
1S/C9H19N3O3.ClH/c1-6(11)8(13)12-7(9(14)15)4-2-3-5-10;/h6-7H,2-5,10-11H2,1H3,(H,12,13)(H,14,15);1H
InChI key
JWNUXAHYKKBLRA-UHFFFAOYSA-N
Biochem/physiol Actions
Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A2003-250MG | 04061832883281 |

