Skip to Content
Merck
All Photos(3)

Documents

A1755

Sigma-Aldrich

Aminophylline

≥98%, powder

Synonym(s):

(Theophylline)2 · Ethylenediamine, 3,7-Dihydro-1,3-dimethyl-1H-purine-2,6-dione, cmpd. with 1,2-ethanediamine (2:1), Theophylline hemi(ethylenediamine) complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2 · 0.5C2H8N2
CAS Number:
Molecular Weight:
210.21
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL (solutions should be freshly prepared.)
alcohol: insoluble

storage temp.

−20°C

SMILES string

NCCN.CN1C(=O)N(C)c2nc[nH]c2C1=O.CN3C(=O)N(C)c4nc[nH]c4C3=O

InChI

1S/2C7H8N4O2.C2H8N2/c2*1-10-5-4(8-3-9-5)6(12)11(2)7(10)13;3-1-2-4/h2*3H,1-2H3,(H,8,9);1-4H2

InChI key

FQPFAHBPWDRTLU-UHFFFAOYSA-N

Application

Aminophylline has been used:
  • as a positive control in the selective inhibitor of nuclear export, KPT-350 zebrafish dosing experiments
  • as a positive control for the determination of the spasmolytic activity of C. aeruginosa extracts
  • as a phosphodiesterase (PDE) inhibitor in inhibitor studies to quantify its potency to inhibit PDE4A1A and PDE3A enzyme in an assay

Biochem/physiol Actions

Aminophylline is a methylxanthine compound containing theophylline andethylenediamine in a ratio of 2:1. It acts as a bronchodilator and is used to treat chronic lung diseases like asthma, bronchitis, or emphysema.
Non-selective phosphodiesterase (PDE) inhibitor. Antiepileptic drug

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danieli Barino Salinas et al.
Pediatric research, 87(1), 137-145 (2019-07-26)
There is a need to prognosticate the severity of cystic fibrosis (CF) detected by newborn screening (NBS) by early assessment of CF transmembrane conductance regulator (CFTR) protein function. We introduce novel instrumentation and protocol for evaluating CFTR activity as reflected
Antiasthmatic effect of Curcuma aeruginosa extract on isolated organ of the trachea
Marliana E, et al.
F1000Research (2018)
Matt Staeben et al.
Assay and drug development technologies, 8(3), 344-355 (2010-02-18)
Screening of AMP- and GMP-producing enzymes such as phosphodiesterases (PDEs), ligases, and synthetases would be simplified by the ability to directly detect unmodified nucleoside monophosphates. To address this need, we developed polyclonal and monoclonal antibodies that recognize AMP and GMP
Thieu X Phan et al.
The Journal of physiology, 598(24), 5639-5659 (2020-09-19)
The functional roles of the capsaicin receptor, TRPV1, outside of sensory nerves are unclear. We mapped TRPV1 in the mouse circulation, revealing extensive expression in the smooth muscle of resistance arterioles supplying skeletal muscle, heart and adipose tissue.  Activation of
Nicole M Bhave et al.
Journal of cardiovascular magnetic resonance : official journal of the Society for Cardiovascular Magnetic Resonance, 14, 89-89 (2013-01-01)
Adenosine cardiovascular magnetic resonance (CMR) can accurately quantify myocardial perfusion reserve. While regadenoson is increasingly employed due to ease of use, imaging protocols have not been standardized. We sought to determine the optimal regadenoson CMR protocol for quantifying myocardial perfusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service