Skip to Content
Merck
All Photos(1)

Documents

A1471

Sigma-Aldrich

A-85783

Synonym(s):

(R)-6-(4-Fluorophenyl)-N,N-dimethyl-3-[3-(3-pyridinyl)-1H,3H-pyrrolo[1,2-c]thiazol-7-yl]carbonyl-1H-indole-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H23FN4O2S
CAS Number:
Molecular Weight:
510.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

mp

185.3-186.5 °C (lit.)

solubility

DMSO: 13 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CN(C)C(=O)n1cc(C(=O)c2ccn3[C@H](SCc23)c4cccnc4)c5ccc(cc15)-c6ccc(F)cc6

InChI

1S/C29H23FN4O2S/c1-32(2)29(36)34-16-24(22-10-7-19(14-25(22)34)18-5-8-21(30)9-6-18)27(35)23-11-13-33-26(23)17-37-28(33)20-4-3-12-31-15-20/h3-16,28H,17H2,1-2H3/t28-/m1/s1

InChI key

YGUVFPOGHIKVPP-MUUNZHRXSA-N

General description

Shown to be effective both in vivo and in vitro. A-85783 is neutral and lipophilic and has been shown to competitively and reversibly inhibit [3H]PAF binding to rabbit platelet membranes as well as block PAF-induced serotonin release from rabbit platelets.

Biochem/physiol Actions

Potent and selective platelet-activating factor (PAF) receptor antagonist.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold under license from Abbott Laboratories.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service