Skip to Content
Merck
All Photos(1)

Key Documents

A1349

Sigma-Aldrich

Monoclonal Anti-β-Amyloid (17-24) antibody produced in mouse

~1 mg/mL, clone 4G8, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

4G8, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:1000
immunoprecipitation (IP): 1:10-1:100
indirect ELISA: 1:103-1:105
western blot: 1:100-1:1000

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APP(351)

Immunogen

synthetic peptide corresponding to amino acids 17-24 of the human β amyloid peptide with Glu substituted at position 11, conjugated to KLH.

Physical form

Solution in phosphate buffered saline.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina Hategan et al.
Nature structural & molecular biology, 24(4), 379-386 (2017-02-22)
Deposition of amyloid-β plaques is increased in the brains of HIV-infected individuals, and the HIV transactivator of transcription (Tat) protein affects amyloidogenesis through several indirect mechanisms. Here, we investigated direct interactions between Tat and amyloid-β peptide. Our in vitro studies
Chunyi Jiang et al.
Analytical and bioanalytical chemistry, 396(5), 1745-1754 (2010-02-06)
Aβ(1-42) is the proteolytic cleavage product of cleavage of the amyloid precursor protein by β- and γ-secretases. The aggregation of Aβ(1-42) plays a causative role in the development of Alzheimer's disease. To lock Aβ(1-42) in a homogenous state, we embedded
M Medecigo et al.
Journal of neuroimmunology, 223(1-2), 104-114 (2010-05-11)
Anti-amyloid immunotherapy has been proposed as an appropriate therapeutic approach for Alzheimer's disease (AD). Significant efforts have been made towards the generation and assessment of antibody-based reagents capable of preventing and clearing amyloid aggregates as well as preventing their synaptotoxic
Stefania Zappettini et al.
Frontiers in pharmacology, 3, 146-146 (2012-08-07)
Using both in vitro (hippocampal synaptosomes in superfusion) and in vivo (microdialysis) approaches we investigated whether and to what extent β amyloid peptide 1-40 (Aβ 1-40) interferes with the cholinergic modulation of the release of glycine (GLY) in the rat
Jason H Y Yeung et al.
Frontiers in neuroscience, 13, 1427-1427 (2020-02-06)
Alzheimer's disease (AD) is the leading type of dementia worldwide. Despite an increasing burden of disease due to a rapidly aging population, there is still a lack of complete understanding of the precise pathological mechanisms which drive its progression. Glutamate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service