Skip to Content
Merck
All Photos(1)

Key Documents

A126

Sigma-Aldrich

(±)-HA-966

Synonym(s):

(±)-3-Amino-1-hydroxy-2-pyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

NC1CCN(O)C1=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valéria Martinez et al.
Pain, 99(3), 537-545 (2002-10-31)
Systemic opioid dosing until adequate analgesia in neuropathic pain may involve intolerable and untreatable side effects. Peripheral opioid receptor mechanisms may participate in the antinociceptive effect of systemic morphine. We evaluated the effect of peripherally injected morphine alone, and the
J Klein et al.
Pharmacopsychiatry, 36 Suppl 1, S78-S83 (2003-09-18)
We have previously shown that hypoxia and N-methyl-D-aspartate (NMDA) receptor activation induce breakdown of choline-containing phospholipids in rat hippocampus, a process which is mediated by calcium influx and phospholipase A (2) activation. Bilobalide, a constituent of Ginkgo biloba, inhibited this
Frédéric Adam et al.
Neuropharmacology, 51(2), 191-202 (2006-05-23)
N-methyl-D-aspartate (NMDA) receptors are widely involved in opioid tolerance. However, it is less clear whether NMDA receptor antagonists reverse already-established tolerance and whether the intensity of the nociceptive stimulus influences morphine tolerance. Three days after implantation of morphine or control
Joanne E Nash et al.
Movement disorders : official journal of the Movement Disorder Society, 17(3), 455-466 (2002-07-12)
Treatments for Parkinson's disease based on replacement of lost dopamine have several problems. Following loss of dopamine, enhanced N-methyl-D-aspartate (NMDA) receptor-mediated transmission in the striatum is thought to be part of the cascade of events leading to the generation of
David L Walker et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(6), 2343-2351 (2002-03-16)
NMDA receptor antagonists block conditioned fear extinction when injected systemically and also when infused directly into the amygdala. Here we evaluate the ability of D-cycloserine (DCS), a partial agonist at the strychnine-insensitive glycine-recognition site on the NMDA receptor complex, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service