Skip to Content
Merck
All Photos(1)

Key Documents

A0476

Sigma-Aldrich

Activin Receptor-Like Kinase 1/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Synonym(s):

ALK-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€652.00

€652.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€652.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

€652.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

mol wt

50-55 kDa by SDS-PAGE (reducing)
calculated mol wt ~37.3 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ACVRL1(94)

Biochem/physiol Actions

ALK-1 is a type I complex of the serine/threonine kinases. Type I and II receptors bind ligands in the TGF-β superfamily. Human and mouse ALK-1 share approximately 71% amino acid sequence identity in their extracellular domains.
Type I complex of the serine/threonine kinases; bind ligands in the TGF-β superfamily.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

Can be used as a calibrator for ALK-1 antibodies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P ten Dijke et al.
Oncogene, 8(10), 2879-2887 (1993-10-01)
Human cDNA clones encoding four novel putative transmembrane protein serine/threonine kinases, denoted activin receptor-like kinase (ALK) -1, -2, -3 and -4, were obtained using a polymerase chain reaction (PCR)-based strategy. The PCR primers were designed based upon the sequence similarity
A Lux et al.
The Journal of biological chemistry, 274(15), 9984-9992 (1999-04-03)
Germ line mutations in one of two distinct genes, endoglin or ALK-1, cause hereditary hemorrhagic telangiectasia (HHT), an autosomal dominant disorder of localized angiodysplasia. Both genes encode endothelial cell receptors for the transforming growth factor beta (TGF-beta) ligand superfamily. Endoglin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service