Skip to Content
Merck
All Photos(1)

Key Documents

95608

Sigma-Aldrich

Lipase from Pseudomonas fluorescens

powder, slightly beige, ~40 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

form

powder

specific activity

~40 U/mg

color

slightly beige

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lipase from Pseudomonas fluorescens has been used to study the activity of lipase in water-restricted environments [1] and has been used to study lipase inactivation [2].

Biochem/physiol Actions

Lipase is an esterase that catalyzes the hydrolysis of ester bonds of lipids. Lipases are essential for the digestion, transport and processing of lipids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314 as substrate)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Soliva-Fortuny et al.
Journal of dairy science, 89(11), 4096-4104 (2006-10-13)
The inactivation kinetics of a lipase from Pseudomonas fluorescens (EC 3.1.1.3.) were studied in a simulated skim milk ultrafiltrate treated with high-intensity pulsed electric fields. Samples were subjected to electric field intensities ranging from 16.4 to 27.4 kV/cm for up
Y Shabtai et al.
Applied and environmental microbiology, 58(1), 174-180 (1992-01-01)
An extracellular lipase from the low-water-tolerant bacterium P. aeruginosa YS-7 was produced, purified, and characterized with respect to its functional properties in aqueous solutions and organic solvents. The enzyme was partially released from the cells during fermentation in defined medium
P L Okorokov et al.
Fiziologiia cheloveka, 38(6), 105-112 (2013-02-12)
Food behavior disorderes (high fat diet) and systematic strong drinks use could be the most important causes of excess intake of intestinal endotoxin (ET) in circulation (endotoxin aggression development) and induction of inflammation. In this work we investigated antiendotoxin immunity
Mehran Howaizi et al.
Acta gastro-enterologica Belgica, 75(4), 446-447 (2013-02-14)
Acute pancreatitis has a large number of causes. Major causes are alcohol and gallstones. Toxic causes, mainly represented by medication-induced pancreatitis account for less than 2% of the cases. Cannabis is an anecdotally reported cause of acute pancreatitis. Six cases
J Hall et al.
The Journal of small animal practice, 54(5), 275-279 (2013-02-13)
Mushroom toxicosis is rarely diagnosed in dogs and is poorly reported in the veterinary literature. This report suggests that mushroom toxicosis is a potentially under-diagnosed condition in first opinion practice in the UK. Nine dogs with clinical signs consistent with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service