Skip to Content
Merck
All Photos(1)

Documents

94261

Sigma-Aldrich

LL-37 (human) trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Human LL-37 amide Trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C205H341N61O52 · xC2HF3O2
Molecular Weight:
4492.28 (free base basis)
UNSPSC Code:
51101500
NACRES:
NA.85

Assay

≥95% (HPLC)

form

powder (Lyophilisate)

color

colorless to white

solubility

H2O: 1 mg/mL, clear, colorless

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
viruses

Mode of action

cell membrane | interferes

storage temp.

−20°C

Related Categories

Amino Acid Sequence

Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser-NH2

General description

Chemical structure: peptide

Application

LL-37 is used to study host defense mechanisms.

Biochem/physiol Actions

LL-37 is a muntifunctional host defense peptide with antibacterial, antiviral, and immunomodulating activities. It is the only cathelicidin-type peptide found in human. In addition to its antimicrobial activities, LL-37 has been found to regulate inflammation and neutralize lipopolysaccharides from Gram-negative bacteria.

Packaging

1mg

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ainhoa Revilla-Guarinos et al.
Applied and environmental microbiology, 86(14) (2020-05-18)
Bce-like systems mediate resistance against antimicrobial peptides in Firmicutes bacteria. Lactobacillus casei BL23 encodes an "orphan" ABC transporter that, based on homology to BceAB-like systems, was proposed to contribute to antimicrobial peptide resistance. A mutant lacking the permease subunit was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service