Skip to Content
Merck
All Photos(1)

Key Documents

921P

Sigma-Aldrich

Declere®

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€92.00
200 ML
€198.00

€92.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€92.00
200 ML
€198.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

€92.00


Please contact Customer Service for Availability

Request a Bulk Order

form

aqueous solution (green)

Quality Level

packaging

pkg of 1 L 20x (921P-09)
pkg of 200 mL 20x (921P-06)
pkg of 50 mL 20x (921P-04)

technique(s)

immunohistochemistry: suitable

shipped in

ambient

General description

Declere may be used whenever immunohistochemistry staining is performed on formalin-fixed, paraffin-embedded tissue sections. Using this product encourages standardization of the pretreatment procedure, thereby producing more consistent, more reliable results.

Physical form

Solution containing pretreatment reagents

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Declere is a registered trademark of CellMarque Corporation

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service