Skip to Content
Merck
All Photos(1)

Documents

90771

Sigma-Aldrich

Poly(maleic anhydride-alt-1-tetradecene), 3-(dimethylamino)-1-propylamine derivative

BioReagent, for molecular biology

Synonym(s):

PMAL®-C12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C23H44O3N2)n
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.25

grade

for molecular biology

description

zwitterionic

product line

BioReagent

form

solid

λ

0.1 % in H2O

UV absorption

λ: 260 nm Amax: 0.400
λ: 280 nm Amax: 0.150

storage temp.

−20°C

Application

Use of "Amphipols" for delivering of membrane proteins to lipid bilayers, as activity support of membrane enzymes.

Analysis Note

enzyme traces DNases, RNases, and phosphatases not detectable

Legal Information

PMAL is a registered trademark of Affymetrix, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bonnie M Gorzelle et al.
Journal of the American Chemical Society, 124(39), 11594-11595 (2002-09-26)
Amphipathic polymers ("amphipols") were introduced several years ago (Tribet, C.; Audebert, R.; Popot, J.-L. Proc. Natl. Acad. Sci. U.S.A. 1996, 93, 15047-15050) as an alternative method for solubilizing integral membrane proteins in stable, nativelike conformations. However, direct maintenance of full
J K Nagy et al.
FEBS letters, 501(2-3), 115-120 (2001-07-27)
Data are presented which suggest that a class of amphiphilic polymers known as 'amphipols' may serve as a vehicle for delivering complex integral membrane proteins into membranes. The integral membrane protein diacylglycerol kinase (DAGK) was maintained in soluble form by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service