90746
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 36mM − Spermine 2.25 mM − PEG 400 5% solution
Synonym(s):
Crystallization solution 6/Kit-No 80701
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
liquid
pH
6.5±0.2 ( in neat)
density
1.02 g/mL at 20 °C
storage temp.
2-8°C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service