Skip to Content
Merck
All Photos(1)

Key Documents

87961

Sigma-Aldrich

Tetranitroblue tetrazolium chloride

≥85% (AT)

Synonym(s):

3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2,5-bis(p-nitrophenyl)-2H-tetrazolium chloride], TNBT, Tetranitrotetrazolium blue chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H28Cl2N12O10
CAS Number:
Molecular Weight:
907.63
Beilstein:
3897475
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥85% (AT)

form

powder or crystals

ign. residue

≤0.1%

loss

≤25% loss on drying

mp

~170 °C (dec.)

solubility

2-methoxyethanol: 0.090 g/10 mL (hot)

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccc(cc4)[N+]([O-])=O)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccc(cc8)[N+]([O-])=O

InChI

1S/C40H28N12O10.2ClH/c1-61-37-23-27(7-21-35(37)47-43-39(25-3-9-31(10-4-25)49(53)54)41-45(47)29-13-17-33(18-14-29)51(57)58)28-8-22-36(38(24-28)62-2)48-44-40(26-5-11-32(12-6-26)50(55)56)42-46(48)30-15-19-34(20-16-30)52(59)60;;/h3-24H,1-2H3;2*1H/q+2;;/p-2

InChI key

VCESGVLABVSDRO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

crystallized with ~2 mol ethanol

Other Notes

Review: Tetrazolium salts and formazans[1]; Detection of hydrolases (with indoxyl substrates)[2]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

F.P. Altman
Progress in Histochemistry and Cytochemistry, 9, 51-51 (1976)
R Gossrau
Histochemistry, 58(3), 203-218 (1978-12-01)
Using freeze-dried or sections from fresh-frozen or aldehyde-fixed material nitro BT (NBT), tetranito BT (TNBT), distyryl nitro BT (DS-NBT), thiocarbamyl nitro BT (TC-NBT) or benzothiazolylstyrylphthalhydrazidyl tetrazolium chloride (BSPT) were tested as auxiliary reagents for the localization of glycosidases, phosphatases and
R L Gardner
Journal of embryology and experimental morphology, 80, 251-288 (1984-04-01)
Conditions were found for staining whole mid-gestation capsular parietal endoderms and visceral yolk sacs for malic enzyme activity that gave excellent discrimination between wild-type (Mod-1+/Mod-1+) cells and mutant (Mod-1n/Mod-1n) cells that lack the cytoplasmic form of the enzyme. Reciprocal blastocyst
W M Frederiks et al.
The Histochemical journal, 25(1), 86-91 (1993-01-01)
A quantitative histochemical procedure was developed for the demonstration of purine nucleoside phosphorylase in rat liver using unfixed cryostat sections and the auxiliary enzyme xanthine oxidase. The optimum incubation medium contained 18% (w/v) poly(vinyl alcohol), 100 mM phosphate buffer, pH
C L Jordan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 9(1), 239-247 (1989-01-01)
In the previous study (Jordan et al., 1989), we demonstrated that androgen treatment of juvenile male rats inhibits the elimination of synapses in the levator ani (LA) muscle. In the present study, we asked whether synapse elimination would occur once

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service