Skip to Content
Merck
All Photos(1)

Key Documents

81460

Sigma-Aldrich

Ponceau S

for microscopy (Hist.)

Synonym(s):

3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
Colour Index Number:
27195
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Pricing and availability is not currently available.

grade

for microscopy (Hist.)

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Nicklas et al.
PloS one, 7(1), e30445-e30445 (2012-02-03)
Limb girdle muscular dystrophy type 2H (LGMD2H) is an inherited autosomal recessive disease of skeletal muscle caused by a mutation in the TRIM32 gene. Currently its pathogenesis is entirely unclear. Typically the regeneration process of adult skeletal muscle during growth
R B DeMattos et al.
Journal of lipid research, 42(6), 976-987 (2001-05-23)
Susceptibility to the development of late-onset Alzheimer's disease is increased for individuals harboring one or more apolipoprotein E4 (apoE4) alleles. Although several isoform-specific effects of apoE have been identified, the relationship between biochemical function and risk factor assessment is unknown.
John C Dreixler et al.
Experimental eye research, 88(3), 512-521 (2008-12-17)
Potent endogenous protection from ischemia can be induced in the retina by ischemic preconditioning (IPC). Protein kinase B/Akt is a cellular survival factor. We hypothesized that Akt was integral to IPC based upon differential effects of Akt subtypes. Rats were
Hong-Wen Gao et al.
Biophysical journal, 94(3), 906-917 (2007-10-02)
Using various methods, e.g., spectrophotometry, circular dichroism, and isothermal titration calorimetry, the interaction of poncean S (PS) with human serum albumin (HSA) was characterized at pH 1.81, 3.56, and 7.40 using the spectral correction technique, and Langmuir and Temkin isothermal
T G van Kooten et al.
Journal of biomedical materials research, 54(3), 385-389 (2001-02-24)
Research of cell-biomaterial interactions is building on knowledge and methods available in cell and molecular biology. Western blotting is one of the options to characterize protein expression in cell populations. Method transfer to biomaterial model systems is not trivial because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service