78896
1,2-Didocosahexaenoyl-sn-glycero-3-phosphocholine
≥99.0% (TLC)
Synonym(s):
PC, (7R,13Z,16Z,19Z,22Z,25Z,28Z)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(4Z,7Z,10Z,13Z,16Z,19Z)-1-oxo-4,7,10,13,16,19-docosahexaenyl]oxy]-3,5,9-trioxa-4-phosphahentriaconta-13,16,19,22,25,28-hexaen-1-aminium 4-oxide, inner salt, 1,2-Di-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine, DHAPC, PC(22:6(4Z,7Z,10Z,13Z,16Z,19Z)/22:6(4Z,7Z,10Z,13Z,16Z,19Z)), PC(22:6/22:6)
About This Item
biological source
synthetic
Assay
≥99.0% (TLC)
form
paste
functional group
phospholipid
storage temp.
−20°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)=O
InChI
1S/C52H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)58-48-50(49-60-62(56,57)59-47-46-53(3,4)5)61-52(55)45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h8-11,14-17,20-23,26-29,32-35,38-41,50H,6-7,12-13,18-19,24-25,30-31,36-37,42-49H2,1-5H3/b10-8-,11-9-,16-14-,17-15-,22-20-,23-21-,28-26-,29-27-,34-32-,35-33-,40-38-,41-39-/t50-/m1/s1
InChI key
XLKQWAMTMYIQMG-SVUPRYTISA-N
General description
Packaging
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service