Skip to Content
Merck
All Photos(1)

Key Documents

78363

Sigma-Aldrich

Atto 490LS maleimide

suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€362.00

€362.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€362.00

About This Item

UNSPSC Code:
12352111
NACRES:
NA.32

€362.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥90% (HPLC)

Quality Level

form

(Report result)

mol wt

Mw 818 g/mol

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm, >90%
490 nm, >90%

fluorescence

λex 496 nm; λem 640.0 nm±10 nm in PBS, pH 7.4

λ

in PBS, pH 7.4

UV absorption

λ: 494-500 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments, in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.
Maleimides are well suited for coupling to thiol groups. This is similar to iodacetamides, but maleimides do react more thiol selective. They do not show significant reaction with histidine or methionine. Hydrolysis of maleimides to a mixture of isomeric nonreactive maleamic acids can compete significantly with thiol modification, particularly above pH 8. Maleimides may be used for labelling of amines, which usually requires a higher pH than reaction of maleimides with thiols.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service