Skip to Content
Merck
All Photos(1)

Key Documents

74908

Sigma-Aldrich

[5-(2-Propynylamino)-9H-benzo[a]phenoxazin-9-ylidene]dimethylammonium chloride

for click labeling, ≥90.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18ClN3O
CAS Number:
Molecular Weight:
363.84
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90.0% (TLC)

form

solid

solubility

methanol: 0.5 mg/mL, clear, blue

fluorescence

λex 633 nm±5 nm; λem 671 nm±10 nm in methanol

storage temp.

−20°C

SMILES string

[Cl-].C\[N+](C)=C1/C=CC2=Nc3c(OC2=C1)cc(NCC#C)c4ccccc34

InChI

1S/C21H17N3O.ClH/c1-4-11-22-18-13-20-21(16-8-6-5-7-15(16)18)23-17-10-9-14(24(2)3)12-19(17)25-20;/h1,5-10,12-13H,11H2,2-3H3;1H

InChI key

HDBXFMBKCWAQCT-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service