Skip to Content
Merck
All Photos(1)

Key Documents

73711

Sigma-Aldrich

2,6-Difluorophenyl pentafluorophenyl decanedioate

≥97.0% (HPLC)

Synonym(s):

Decanedioic acid 1-(2,6-difluorophenyl) 10-(2,3,4,5,6-pentafluorophenyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19F7O4
CAS Number:
Molecular Weight:
480.37
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28
Pricing and availability is not currently available.

Assay

≥97.0% (HPLC)

form

lumps

SMILES string

O=C(CCCCCCCCC(OC1=C(F)C=CC=C1F)=O)OC2=C(F)C(F)=C(F)C(F)=C2F

InChI

1S/C22H19F7O4/c23-12-8-7-9-13(24)21(12)32-14(30)10-5-3-1-2-4-6-11-15(31)33-22-19(28)17(26)16(25)18(27)20(22)29/h7-9H,1-6,10-11H2

InChI key

UVKRJTSPLZDUFT-UHFFFAOYSA-N

Application

This linker enables to form heteroconjugates by linking primary amino groups from two separate biomolecules in a strictly orthogonal way. The orthogonality arises from the differential reactivity of the ester leaving groups.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam T Slósarczyk et al.
Journal of peptide science : an official publication of the European Peptide Society, 18(4), 261-269 (2012-03-07)
An efficient method for the heteroconjugation of biomolecules carrying free amino groups was reported previously, where mixed polyfluorophenyl diesters of dicarboxylic acids with varied aliphatic chain length were shown to be efficient reagents for the conjugation of a variety of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service