Skip to Content
Merck
All Photos(1)

Documents

73499

Sigma-Aldrich

Atto MB2 NHS ester

Methylene Blue derivative, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

Assay

≥90% (HPLC)
≥90% (degree of coupling)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in acetonitrile (with 0.1% acetic acid)

UV absorption

λ: 652-658 nm Amax

storage temp.

−20°C

General description

Atto MB2 NHS ester is a derivative of the well-known redox dye Methylene Blue. The dye can be reversibly reduced to the colorless leuko form. Upon oxidation (e.g. with oxygen) the dye recovers, and the absorption is fully restored.
The dye is suitable for labeling of DNA, RNA, proteins etc. In common with most Atto-labels the dye shows a high extinction coefficient.
Atto MB2 NHS ester is moderately hydrophilic.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service