Skip to Content
Merck
All Photos(1)

Key Documents

73494

Sigma-Aldrich

Dy-560 NHS ester

for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H44N4O10S2
CAS Number:
Molecular Weight:
768.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Quality Level

form

solid

fluorescence

λex 568 nm; λem 588 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for coupling to amines

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2c(-c3ccc(cc3S([O-])(=O)=O)S(=O)(=O)NCCCCCC(=O)ON4C(=O)CCC4=O)c5ccc(cc5[o+]c2c1)N(CC)CC

InChI

1S/C37H44N4O10S2/c1-5-39(6-2)25-13-16-28-31(22-25)50-32-23-26(40(7-3)8-4)14-17-29(32)37(28)30-18-15-27(24-33(30)53(47,48)49)52(45,46)38-21-11-9-10-12-36(44)51-41-34(42)19-20-35(41)43/h13-18,22-24,38H,5-12,19-21H2,1-4H3

InChI key

ZVRXEZSQXWSLKP-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service