Skip to Content
Merck
All Photos(1)

Key Documents

73361

Sigma-Aldrich

Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulfone

≥97.0% (CHN)

Synonym(s):

BSOCOES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O12S
CAS Number:
Molecular Weight:
436.35
Beilstein:
1521053
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0% (CHN)

reaction suitability

reagent type: cross-linking reagent

mp

169-173 °C

storage temp.

−20°C

SMILES string

O=C1CCC(=O)N1OC(=O)OCCS(=O)(=O)CCOC(=O)ON2C(=O)CCC2=O

InChI

1S/C14H16N2O12S/c17-9-1-2-10(18)15(9)27-13(21)25-5-7-29(23,24)8-6-26-14(22)28-16-11(19)3-4-12(16)20/h1-8H2

InChI key

XUDGDVPXDYGCTG-UHFFFAOYSA-N

Other Notes

Homobifunctional crosslinking reagent for proteins; cross-linking reagents used as molecular rulers

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Antonian et al.
European journal of biochemistry, 145(3), 511-517 (1984-12-17)
Calf brain plain synaptic vesicle proteins have been cross-linked with bis[2-(succinimidooxycarbonyloxy)ethyl] sulfone, a homobifunctional, cleavable reagent, as well as with N-hydroxysuccinimidyl 4-azidobenzoate, a photosensitive, heterobifunctional reagent. These results demonstrate the generality of a recent report that synaptic vesicle proteins can
J Shin et al.
The Journal of biological chemistry, 260(23), 12822-12827 (1985-10-15)
Both of the alpha and beta subunits of intact human follitropin (FSH) were radioiodinated with 125I-sodium iodide and chloramine-T and could be resolved on sodium dodecyl sulfate-polyacrylamide gels. Radioiodinated FSH was affinity-cross-linked with a cleavable (nondisulfide) homobifunctional reagent to its
R A Smith et al.
The Journal of biological chemistry, 261(21), 9850-9853 (1986-07-25)
The quaternary structural relationships between subunits of the follitropin (FSH) receptor were determined through the use of the reversible, homobifunctional, chemical cross-linking reagent bis[2-(succinimidooxycarbonyloxy)ethyl]sulfone (BSOCOES) after formation of covalent 125I-azidobenzoyl-FSH-receptor subunit complexes by photoaffinity labeling. This experimental approach resulted in
Z Bouizar et al.
European journal of biochemistry, 155(1), 141-147 (1986-02-17)
We have characterized the binding parameters of renal receptors (Scatchard analysis revealed the presence of two binding sites: site I, Ka1 = 1.29 X 10(9) M-1, number of binding sites = 9.9 X 10(6)/micrograms protein; site II, Ka2 = 0.93
P Gourlet et al.
Journal of receptor research, 11(5), 831-848 (1991-01-01)
Cross-linking of [125I]helodermin to human SUP-T1 lymphoblasts with bis[2-(succinimidooxycarbonyloxy)ethyl]sulfone (BSOCOES) revealed a 63 K binding protein. This cross-linking was inhibited by helodermin and VIP. In cells submitted for 3-4 days to 0.2 microgram/ml tunicamycin, the Mr of an increasing proportion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service