Skip to Content
Merck
All Photos(2)

Key Documents

71770

Sigma-Aldrich

4-Nitrophenyl phosphate disodium salt hexahydrate

≥97.0% (NT)

Synonym(s):

p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
Beilstein:
5324661
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0% (NT)

form

powder or crystals

impurities

≤0.01% free nitrophenol

solubility

H2O: 25 mg/mL at 20 °C, clear, yellow-green

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chromogenic substrate for the determination of acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Papista et al.
Kidney international, 88(2), 276-285 (2015-03-26)
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the
Masaya Yokota et al.
Arthritis research & therapy, 16(2), R72-R72 (2014-03-19)
In addition to the pivotal roles of mast cells in allergic diseases, recent data suggest that mast cells play crucial roles in a variety of autoimmune responses. However, their roles in the pathogenesis of autoimmune skeletal muscle diseases have not
Rasmus Iversen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), 17146-17151 (2014-11-19)
The multifunctional enzyme transglutaminase 2 (TG2) is the target of autoantibodies in the gluten-sensitive enteropathy celiac disease. In addition, the enzyme is responsible for deamidation of gluten peptides, which are subsequently targeted by T cells. To understand the regulation of
Jörn Karhausen et al.
Critical care medicine, 41(9), e200-e210 (2013-03-13)
Cardiac surgery, especially when employing cardiopulmonary bypass and deep hypothermic circulatory arrest, is associated with systemic inflammatory responses that significantly affect morbidity and mortality. Intestinal perfusion abnormalities have been implicated in such responses, but the mechanisms linking local injury and
Erich V Scheller et al.
mBio, 6(3), e00500-e00515 (2015-05-28)
Bordetella fimbriae (FIM) are generally considered to function as adhesins despite a lack of experimental evidence supporting this conclusion for Bordetella pertussis and evidence against a requirement for FIM in adherence of Bordetella bronchiseptica to mammalian cell lines. Using B. bronchiseptica

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service