Skip to Content
Merck
All Photos(1)

Documents

68854

Sigma-Aldrich

L-Arabino-1,4-lactone

≥95.0% (GC)

Synonym(s):

D-Arabinonic acid γ-lactone, L-Arabinono-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
Beilstein:
82060
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (GC)

form

powder

optical activity

[α]/D -72.0±8.0°, c = 1 in H2O

suitability

conforms to structure for Proton NMR spectrum

SMILES string

O=C1[C@H](O)[C@@H](O)[C@H](CO)O1

InChI

1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4+/m0/s1

InChI key

CUOKHACJLGPRHD-YVZJFKFKSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of Hemicellulose from Loblolly Pine Woodchips and Subsequent Kraft Pulping.
Huang, F. and Ragauskas, A.
Industrial & Engineering Chemistry Research, 52, 1743-1749 (2013)
Robert M Archer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(8), 2895-2902 (2013-01-15)
Practical syntheses of 2-keto-3-deoxy-D-xylonate (D-KDX) and 2-keto-3-deoxy-L-arabinonate (L-KDA) that rely on reaction of the anion of ethyl 2-[(tert-butyldimethylsilyl)oxy]-2-(dimethoxy phosphoryl) acetate with enantiopure glyceraldehyde acetonide, followed by global deprotection of the resultant O-silyl-enol esters, have been developed. This has enabled us
Medel Manuel L Zulueta et al.
Chemical communications (Cambridge, England), 49(32), 3275-3287 (2013-02-28)
Carbohydrates either conjugated or as free entities are major players in numerous biological processes. The desire to comprehend the nature of their functions and further develop therapeutic and diagnostic applications has fuelled the recent upsurge in the glycoscience field. Mainly
Dominik Mojzita et al.
FEBS letters, 584(16), 3540-3544 (2010-07-27)
L-Xylulose reductase is part of the eukaryotic pathway for l-arabinose catabolism. A previously identified L-xylulose reductase in Hypocrea jecorina turned out to be not the 'true' one since it was not upregulated during growth on L-arabinose and the deletion strain
Ana S P Moreira et al.
Journal of mass spectrometry : JMS, 49(4), 280-290 (2014-04-11)
The oxidation of α-(1 → 5)-L-arabinotriose (Ara3), an oligosaccharide structurally related to side chains of coffee arabinogalactans, was studied in reaction with hydroxyl radicals generated under conditions of Fenton reaction (Fe(2+)/H2O2). The acidic and neutral oxidation products were separated by ligand exchange/size-exclusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service