Skip to Content
Merck
All Photos(1)

Key Documents

68713

Sigma-Aldrich

D-Xylonic acid ammonium salt

≥95% (HPLC)

Synonym(s):

Ammonium D-xylonate, NSC 15831

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O6 · NH3
CAS Number:
Molecular Weight:
183.16
Beilstein:
3742962
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D 18.5±2.0°, c = 1 in H2O

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.N

InChI

1S/C5H10O6.H3N/c6-1-2(7)3(8)4(9)5(10)11;/h2-4,6-9H,1H2,(H,10,11);1H3/t2-,3+,4-;/m1./s1

InChI key

VUOKJALMVMFERM-HAYYNCRMSA-N

Biochem/physiol Actions

Metabolite for the xylose utilization pathway.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mervi H Toivari et al.
Applied microbiology and biotechnology, 96(1), 1-8 (2012-08-10)
D-Xylonic acid is a versatile platform chemical with reported applications as complexing agent or chelator, in dispersal of concrete, and as a precursor for compounds such as co-polyamides, polyesters, hydrogels and 1,2,4-butanetriol. With increasing glucose prices, D-xylonic acid may provide
Huaiwei Liu et al.
Bioresource technology, 115, 244-248 (2011-09-16)
An engineered Escherichia coli was constructed to produce D-xylonic acid, one of the top 30 high-value chemicals identified by US Department of Energy. The native pathway for D-xylose catabolism in E. coli W3110 was blocked by disrupting xylose isomerase (XI)
Mervi Toivari et al.
Bioresource technology, 133, 555-562 (2013-03-05)
D-xylonic acid is one of the top 30 most desirable chemicals to be derived from biomass sugars identified by the US Department of Energy, being applicable as a non-food substitute for D-gluconic acid and as a platform chemical. We engineered
Note on D-xylonate utilization by ascomycetous and basidiomycetous yeasts.
Van Der Walt, J. P. and Steyn, R. L.
Systematic and Applied Microbiology, 13, 192-193 (1990)
Mervi Toivari et al.
Metabolic engineering, 14(4), 427-436 (2012-06-20)
An NAD(+)-dependent D-xylose dehydrogenase, XylB, from Caulobacter crescentus was expressed in Saccharomyces cerevisiae, resulting in production of 17 ± 2 g D-xylonate l(-1) at 0.23 gl(-1)h(-1) from 23 g D-xylose l(-1) (with glucose and ethanol as co-substrates). D-Xylonate titre and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service