Skip to Content
Merck
All Photos(1)

Documents

68617

Sigma-Aldrich

10-Methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16FNO5S
CAS Number:
Molecular Weight:
413.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[O-]S(F)(=O)=O.C[n+]1c2ccccc2c(C(=O)Oc3ccccc3)c4ccccc14

InChI

1S/C21H16NO2.FHO3S/c1-22-18-13-7-5-11-16(18)20(17-12-6-8-14-19(17)22)21(23)24-15-9-3-2-4-10-15;1-5(2,3)4/h2-14H,1H3;(H,2,3,4)/q+1;/p-1

InChI key

DMGQABMILTYHNY-UHFFFAOYSA-M

Application

10-Methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate has been used in the chemiluminescence reaction for the quantification of hydrogen peroxide.

Biochem/physiol Actions

10-methyl-9-(phenoxycarbonyl) acridinium fluorosulfonate (PMAC) has the ability to produce chemiluminescence under neutral conditions.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Acridinium ester for chemiluminescent immunoassay

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Kawaguchi and N. Suzuki et al.
Proc. 9th Int. Symp. Biolumin. Chemilumin., 481-481 (1997)
Development of a highly sensitive chemiluminescent assay for hydrogen peroxide under neutral conditions using acridinium ester and its application to an enzyme immunoassay
Arakawa H, et al.
Luminescence, 29(4), 374-377 (2014)
Extracellular hydrogen peroxide measurements using a flow injection system in combination with microdialysis probes-Potential and challenges
Mosshammer M, et al.
Free Radicals in Biology, 29(4), 374-377 (2018)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service