Skip to Content
Merck
All Photos(1)

Key Documents

67228

Sigma-Aldrich

Poly(maleic anhydride-alt-1-decene), 3-(dimethylamino)-1-propylamine derivative

BioReagent, for molecular biology

Synonym(s):

PMAL®-C8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C19H36O3N2)n
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.25

biological source

synthetic

grade

Molecular Biology
for molecular biology

description

zwitterionic

product line

BioReagent

color

white to off-white

λ

0.1 % in H2O

UV absorption

λ: 260 nm Amax: ≤0.550
λ: 280 nm Amax: ≤0.200

application(s)

hematology
histology

storage temp.

room temp

Analysis Note

enzyme traces DNases, RNases, proteases and phosphatases not detectable

Legal Information

PMAL is a registered trademark of Affymetrix, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Nagy et al.
FEBS letters, 501(2-3), 115-120 (2001-07-27)
Data are presented which suggest that a class of amphiphilic polymers known as 'amphipols' may serve as a vehicle for delivering complex integral membrane proteins into membranes. The integral membrane protein diacylglycerol kinase (DAGK) was maintained in soluble form by
Bonnie M Gorzelle et al.
Journal of the American Chemical Society, 124(39), 11594-11595 (2002-09-26)
Amphipathic polymers ("amphipols") were introduced several years ago (Tribet, C.; Audebert, R.; Popot, J.-L. Proc. Natl. Acad. Sci. U.S.A. 1996, 93, 15047-15050) as an alternative method for solubilizing integral membrane proteins in stable, nativelike conformations. However, direct maintenance of full

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service