Skip to Content
Merck
All Photos(1)

Key Documents

67190

Sigma-Aldrich

1,2,3-Heptanetriol Isomer T 1 M solution

Synonym(s):

1,2,3-Heptanetriol solution, Additive Screening Solution 09/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CCCCC(O)C(O)CO

InChI

1S/C7H16O3/c1-2-3-4-6(9)7(10)5-8/h6-10H,2-5H2,1H3

InChI key

HXYCHJFUBNTKQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Rosenow et al.
Acta crystallographica. Section D, Biological crystallography, 59(Pt 8), 1422-1428 (2003-07-24)
The effect of detergents and amphiphiles on protein solubility and their use in crystallization solutions was examined for an integral membrane protein, the light-harvesting I complex from Rhodospirillum centenum. Measurement by a centrifugation assay of the solubility of the protein
T Okada et al.
Journal of structural biology, 130(1), 73-80 (2000-05-12)
Rhodopsin, a prototypic G protein-coupled receptor responsible for absorption of photons in retinal rod photoreceptor cells, was selectively extracted from bovine rod outer segment membranes, employing mixed micelles of nonyl beta-d-glucoside and heptanetriol. Highly purified rhodopsin was crystallized from solutions
J P Allen
Proteins, 20(3), 283-286 (1994-11-01)
The reaction center from the nonsulfur purple bacterium Rhodobacter sphaeroides has been crystallized in a new form. The crystals grew in the presence of polyethylene glycol 4000, the detergent beta-octyl glucoside, and the amphiphiles heptane triol and benzamidine hydrochloride, using
Dario Piano et al.
Photosynthesis research, 106(3), 221-226 (2010-11-11)
Photosystem II from transplastomic plants of Nicotiana tabacum with a hexahistidine tag at the N-terminal end of the PsbE subunit (α-chain of the cytochrome b(559)) was purified according to the protocol of Fey et al. (BBA 12:1501-1509, 2008). The protein
P Gast et al.
European journal of biochemistry, 239(3), 805-809 (1996-08-01)
Detergent-free reaction centers from Rhodobacter sphaeroides R26 were used to study the solubilization of reaction centers in various detergents and their effects on reaction center photochemistry. 500 +/- 100 n-octyl-beta-D-glucopyranoside or 51 +/- 5 Triton X-100 molecules were associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service