Skip to Content
Merck
All Photos(1)

Documents

66540632

Sigma-Aldrich

BIONi010-C-17

Human iPS Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509

biological source

human

reprogramming method

episomal

description

age (N/A)

manufacturer/tradename

EBiSC

gender

male

growth mode

adherent (pluripotent)

technique(s)

cell culture | stem cell: suitable

relevant disease(s)

Alzheimer′s disease

shipped in

dry ice

storage temp.

−196°C

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Cell Line Origin

Depositor
Bioneer

Cell Line Description

Derivation
Primary cell type: -
Reprogramming method
Vector type: Non-integrating
Vector: Episomal
Is the reprogramming vector detectable: Unknown

Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no

Characterization

Analysis of Undifferentiated Cells
Morphology:
TREM2 KO cl. 24-54 D1 thaw aft_ekU7aDe.tif - Phase contrast image 1 day after thawing
Differentiation potency
Ectoderm:
Ectoderm
In vitro directed differentiation
Morphology:
BIONi010-C-17 ecto D5 10x.tiff
Protocol or reference:
BIONi010-C-17 ecto D5.tif - Flow cytometry of in vitro differentiation to ectoderm
Endoderm:
Endoderm
In vitro directed differentiation
Morphology:
BIONi010-C-17 endo D5 10x.tiff
Protocol or reference:
BIONi010-C-17 endo D5.tif - Flow cytometry of in vitro differentiation to endoderm
Mesoderm:
Mesoderm
In vitro directed differentiation
Morphology:
BIONi010-C-17 meso D5 10x.tiff
Protocol or reference:
BIONi010-C-17 meso D5.tif - Flow cytometry of in vitro differentiation to mesoderm
Microbiology / Virus Screening
HIV 1: -
HIV 2: -
Hepatitis B: -
Hepatitis C: -
Mycoplasma: Negative

Genotyping
Karyotyping
Passage number: 35
Cell line karyotype: 46, XY
Karyotyping method: G-Banding
Image / Result

Genetic Modification

Disease/phenotype related modifications
Disease: Alzheimer′s disease
Type of modification: Gene knock-out
Gene: TREM2
Chromosome location: KO of exon 1 of TREM2 Transcript variant 1 NM_018965
Delivery method: CRISPR-associated (CRISPR/Cas) System

Linkage

Note: EAUA and CLIP must be completed before order fulfillment

Subculture Routine

Surface coating: Matrigel®/Geltrex®
Passage method: Enzyme-free cell dissociation / EDTA
O2 concentration: 20%
CO2 concentration: 5%
Culture medium: Essential E8®
Passage Number banked (pre-EBiSC): 35
No. Vials banked (pre-EBiSC): 30

Legal Information

E8 is a registered trademark of WIsconsin Alumni Research Foundation non-stock Corporation
EBiSC is a trademark of Fraunhofer-Gesellschaft
GELTREX is a registered trademark of Life Technologies Corporation
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service