Skip to Content
Merck
All Photos(2)

Key Documents

64000WB

Midas® Water Bath

new

for use with Midas® III-Plus Automated Stainer

Synonym(s):

Stainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103706

carton L × W × H

26 in. × 16 in. × 7 in.

water bath L × W × H

22 in. × 7.5 in. × 6 in.

compatibility

for use with Midas® III-Plus Automated Stainer

shipped in

ambient

Application

MIDAS®

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service