Skip to Content
Merck
All Photos(1)

Key Documents

62315

Sigma-Aldrich

Lipid Crimson

for microscopy

Synonym(s):

1-(4-[Phenylazo]phenylazo)-2-methylaminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19N5
CAS Number:
Molecular Weight:
365.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for microscopy

SMILES string

CNc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C23H19N5/c1-24-22-16-11-17-7-5-6-10-21(17)23(22)28-27-20-14-12-19(13-15-20)26-25-18-8-3-2-4-9-18/h2-16,24H,1H3/b26-25+,28-27+

InChI key

NVNDCGNXYIBOLU-PAMTUDGESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lipid Crimson is used as a lipoprotein stain for microscopy.

Other Notes

Lipoprotein stain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Raymond et al.
Science (New York, N.Y.), 151(3708), 346-347 (1966-01-21)
Lipoproteins of serum stained before electrophoresis gave distinct patterns in acrylamide gel of 3-percent concentration. Samples compared in parallel on gel slabs showed qualitative differences in migration rates of beta-lipoprotein bands.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service