Skip to Content
Merck
All Photos(1)

Key Documents

56845

Sigma-Aldrich

L-(+)-Erythrulose

≥75% (HPLC)

Synonym(s):

S-1,3,4-Trihydroxy-2-butanone, L-Glycero-2-tetrulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥75% (HPLC)

form

liquid

optical activity

[α]/D 12.0±2.0°, c = 2 in H2O (24 h)

impurities

≤23% water

color

light yellow

storage temp.

room temp

SMILES string

OC[C@H](O)C(=O)CO

InChI

1S/C4H8O4/c5-1-3(7)4(8)2-6/h3,5-7H,1-2H2/t3-/m0/s1

InChI key

UQPHVQVXLPRNCX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-(+)-Erythrulose is used as a tanning agent in the cosmetics industry and a source of chiral ethyl ketones used in aldo reaction organic synthesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santiago Díaz-Oltra et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(30), 9240-9254 (2008-08-30)
Both matched and mismatched diastereoselection have been observed in aldol reactions of a boron enolate of a protected L-erythrulose derivative with several chiral alpha-fluoro and alpha-amino aldehydes. Strict adherence to the Felkin-Anh model for the respective transition structures does not
G L Simpson et al.
Biochimica et biophysica acta, 1501(1), 12-24 (2000-03-23)
The degradation of L-ascorbate (AsA) and its primary oxidation products, L-dehydroascorbate (DHA) and 2,3-L-diketogulonate (2, 3-DKG) were studied under physiological conditions. Analysis determined that L-erythrulose (ERU) and oxalate were the primary degradation products of ASA regardless of which compound was
Xingxing Zou et al.
Journal of agricultural and food chemistry, 65(35), 7721-7725 (2017-07-15)
L-erythrose, a rare aldotetrose, possesses various pharmacological activities. However, efficient L-erythrose production is challenging. Currently, L-erythrose is produced by a two-step fermentation process from erythritol. Here, we describe a novel strategy for the production of L-erythrose in Gluconobacter oxydans (G.
Santiago Díaz-Oltra et al.
The Journal of organic chemistry, 70(20), 8130-8139 (2005-11-10)
[Chemical reaction: See text] Both matched and mismatched diastereoselections have been observed in the aldol reactions of a range of chiral aldehydes with the dicyclohexylboron enolate of a chiral ethyl ketone related to L-erythrulose. As was previously observed in the
Matilte Halma et al.
Biosensors & bioelectronics, 87, 850-857 (2016-09-23)
In the present work, we describe a new thiamine amperometric biosensor based on thiamine pyrophosphate (ThDP)-dependent transketolase (TK)-catalyzed reaction, followed by the oxidative trapping of TK intermediate α,β-dihydroxyethylthiamine diphosphate (DHEThDP) within the enzymatic active site. For the biosensor design purpose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service