Skip to Content
Merck
All Photos(1)

Documents

56783

Sigma-Aldrich

Crystallization Kit for Proteins for Automatic Screening

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

storage temp.

2-8°C

General description

The Automated Screening Kit contains 96 components of 1 mL each in a 8 x 12 deep-well plate. This kit covers most of the solutions contained in the Crystallization Basic Kit (Cat. No. 82009) and Crystallization Extention Kit (Cat. No. 70437).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service