Skip to Content
Merck
All Photos(1)

Key Documents

56592

Sigma-Aldrich

1,2,3-Octanetriol Isomer H 0.1 M solution

Synonym(s):

1,2,3-Octanetriol solution, Additive Screening Solution 10/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

CCCCCC(O)C(O)CO

InChI

1S/C8H18O3/c1-2-3-4-5-7(10)8(11)6-9/h7-11H,2-6H2,1H3

InChI key

QSQGNBGEHSFMAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service