Skip to Content
Merck
All Photos(1)

Key Documents

55156

Sigma-Aldrich

7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester

suitable for fluorescence, ≥95.0% (coupling assay to aminopropyl silica gel)

Synonym(s):

N-Succinimidyl 7-hydroxycoumarin-3-carboxylate, Umbelliferone-3-carboxylic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€206.00
100 MG
€662.00

€206.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€206.00
100 MG
€662.00

About This Item

Empirical Formula (Hill Notation):
C14H9NO7
CAS Number:
Molecular Weight:
303.22
Beilstein:
9419622
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

€206.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95.0% (coupling assay to aminopropyl silica gel)

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 386 nm; λem 448 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

Oc1ccc2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)Oc2c1

InChI

1S/C14H9NO7/c16-8-2-1-7-5-9(13(19)21-10(7)6-8)14(20)22-15-11(17)3-4-12(15)18/h1-2,5-6,16H,3-4H2

InChI key

KFEBWCYYRFZMTJ-UHFFFAOYSA-N

Application

7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester (7-OHCCA-SE) is used as a reagent to conjugate 7-Hydroxycoumarin-3-carboxylic acid (7-OHCCA) to other molecules such as amino acids via amide chemistry.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R Cordfunke et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(13), 7396-7401 (1998-06-24)
The chromophore of photoactive yellow protein (PYP) (i.e., 4-hydroxycinnamic acid) has been replaced by an analogue with a triple bond, rather than a double bond (by using 4-hydroxyphenylpropiolic acid in the reconstitution, yielding hybrid I) and by a "locked" chromophore
Y Oda et al.
Analytical biochemistry, 269(2), 230-235 (1999-05-01)
A fluorometric binding assay for lectin and yeast cells using the avidin-biotin system was previously reported (Y. Oda, M. Kinoshita, and K. Kakehi, Anal. Biochem. 254, 41-48, 1997). However, the true amount of bound lectin could not be determined by

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service