Skip to Content
Merck
All Photos(1)

Key Documents

54297

Sigma-Aldrich

(−)-α-Conidendrin

≥95.0% (HPLC)

Synonym(s):

(3aR,4S,9aR)-3a,4,9,9a-Tetrahydro-6-hydroxy-4-(4-hydroxy-3-methoxyphenyl)-7-methoxynaphtho[2,3-c]furan-1(3H)-one, (3aR,4S,9aR)-6-Hydroxy-4-(4-hydroxy-3-methoxy-phenyl)-7-methoxy-3a,4,9,9a-tetrahydro-3H-naphtho[2,3-c]furan-1-one, NSC 4586, Tsugalactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O6
CAS Number:
Molecular Weight:
356.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (HPLC)

SMILES string

COc1cc2C[C@@H]3[C@H](COC3=O)[C@@H](c4ccc(O)c(OC)c4)c2cc1O

InChI

1S/C20H20O6/c1-24-17-6-10(3-4-15(17)21)19-12-8-16(22)18(25-2)7-11(12)5-13-14(19)9-26-20(13)23/h3-4,6-8,13-14,19,21-22H,5,9H2,1-2H3/t13-,14+,19+/m1/s1

InChI key

CAYMSCGTKZIVTN-TYILLQQXSA-N

Application

α-conidendrin is a plant derived lignan that may be used as a source or reference compound to study bacteria that degrade plant lignans. α-conidendrin may be used as a starting material for the synthesis of sikkimotoxin derivatives and cytotoxic metabolites.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert T LaLonde et al.
Journal of natural products, 67(4), 697-699 (2004-04-24)
Oxidation of alpha-conidendrin (3) by Fremy's salt favored formation of an o-quinone (4) at the pendant aromatic ring as opposed to the fused aromatic ring. Quinone reduction and phenolic methylation, followed by lactone reduction, and subsequent oxidation by dichlorodicyanoquinone produced
Annika I Smeds et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 816(1-2), 87-97 (2005-01-25)
Ten potential lignan metabolites were quantified in rat urine extracts using liquid chromatography-tandem mass spectrometry. The rats were orally administered with the plant lignans 7-hydroxymatairesinol, matairesinol, lariciresinol or secoisolariciresinol, or with the mammalian lignan enterolactone. The samples were enzymatically hydrolysed
Yimin Hu et al.
Chemistry, an Asian journal, 5(2), 309-314 (2009-12-31)
An efficient domino cyclization method for the construction of aza-podophyllotoxin/aza-conidendrin derivatives has been established. Reactions of different dienes with aryl halides in the presence of a palladium catalytic system produced different kinds of podophyllotoxin derivatives through a highly regioselective C-H
A Dantzig et al.
Journal of medicinal chemistry, 44(2), 180-185 (2001-02-15)
Derivatives of alpha-conidendrin, podophyllotoxin, and sikkimotoxin were prepared to evaluate the cytotoxic contributions of C-4 configuration and pendant and fused arene substitutions. Dimethyl-alpha-conidendryl alcohol (5), 9-deoxypodophyllol (6), and 9-deoxysikkimol (17) were dehydrated to their respective oxolane derivatives 4, 3, and
Joshua Fischer et al.
Organic letters, 6(9), 1345-1348 (2004-04-23)
[reaction: see text] Total syntheses of seven biologically important lignan natural products, including (-)-arctigenin, (-)-matairesinol, and (-)-alpha-conidendrin, by way of a highly stereoselective domino radical sequence is presented. The reported stereochemistry of the natural product 7-hydroxyarctigenin is shown to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service